missing translation for 'onlineSavingsMsg'
Learn More

Azetidine hydrochloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 36520-39-5
Molecular Formula C3H7N·HCl
Molecular Weight (g/mol) 93.56
MDL Number MFCD00191762
InChI Key HGQULGDOROIPJN-UHFFFAOYSA-N
Synonym azetidine hydrochloride, azetidine hcl, trimethyleneimine hydrochloride, azetidine, hydrochloride, azetidinehydrochloride, azetidine-hcl, azetidine hcl salt, azetidine-hydrochloride, azetidine; hydrochloride, acmc-1aiho
PubChem CID 12308726
IUPAC Name azetidine;hydrochloride
SMILES C1CNC1.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC398730010
SDS View Documents
Thermo Scientific Chemicals
398730010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC398730050
SDS View Documents
Thermo Scientific Chemicals
398730050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

36520-39-5
93.56
HGQULGDOROIPJN-UHFFFAOYSA-N
12308726
C1CNC1.Cl
C3H7N·HCl
MFCD00191762
azetidine hydrochloride, azetidine hcl, trimethyleneimine hydrochloride, azetidine, hydrochloride, azetidinehydrochloride, azetidine-hcl, azetidine hcl salt, azetidine-hydrochloride, azetidine; hydrochloride, acmc-1aiho
azetidine;hydrochloride

Specifications

36520-39-5
97%
C3H7N·HCl
1g
HGQULGDOROIPJN-UHFFFAOYSA-N
azetidine;hydrochloride
12308726
97%
152°C
Glass bottle
MFCD00191762
azetidine hydrochloride, azetidine hcl, trimethyleneimine hydrochloride, azetidine, hydrochloride, azetidinehydrochloride, azetidine-hcl, azetidine hcl salt, azetidine-hydrochloride, azetidine; hydrochloride, acmc-1aiho
C1CNC1.Cl
93.56
93.56
Azetidine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only