missing translation for 'onlineSavingsMsg'
Learn More

Azetidine-2-carboxylic acid

Catalog No. AC466900050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466900050 5 g
AC466900010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466900050 Supplier Thermo Scientific Chemicals Supplier No. 466900050
Only null left

CAS: 2517-04-6 | C4H7NO2 | 101.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2517-04-6
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
InChI Key IADUEWIQBXOCDZ-UHFFFAOYNA-N
IUPAC Name azetidine-2-carboxylic acid
SMILES OC(=O)C1CCN1

Specifications

Quantity 5 g
Formula Weight 101.1
Chemical Name or Material Azetidine-2-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 219-740-2

RUO – Research Use Only