missing translation for 'onlineSavingsMsg'
Learn More

Azer Scientific Pre-Filled 10% Neutral Buffered Formalin

Catalog No. p-4672658
Change view
Click to view available options
Capacity:
20 mL
40 mL
60 mL
90 mL
120 mL
Content And Storage:
10 mL NBF 10%
20 mL NBF 10%
30 mL NBF 10%
45 mL NBF 10%
60 mL NBF 10%
Packaging Type:
12 Pk., 20 Pks/Cs.
12 Pk., 4 Pks/Cs.
24 Pk., 2 Pks/Cs.
24 Pk., 20 Pks/Cs.
24 Pk., 8 Pks/Cs.
27 Pk., 12 Pks/Cs.
32 Pk., 24 Pks/Cs.
32 Pk., 4 Pks/Cs.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Content And Storage Packaging Type
22026435 20 mL 10 mL NBF 10% 32 Pk., 4 Pks/Cs.
22026436 20 mL 10 mL NBF 10% 32 Pk., 24 Pks/Cs.
22026437 40 mL 20 mL NBF 10% 24 Pk., 2 Pks/Cs.
22026438 40 mL 20 mL NBF 10% 24 Pk., 20 Pks/Cs.
22026440 60 mL 30 mL NBF 10% 27 Pk., 12 Pks/Cs.
22026441 90 mL 45 mL NBF 10% 12 Pk., 4 Pks/Cs.
22026442 90 mL 45 mL NBF 10% 12 Pk., 20 Pks/Cs.
22026443 120 mL 60 mL NBF 10% 24 Pk., 2 Pks/Cs.
22026444 120 mL 60 mL NBF 10% 24 Pk., 8 Pks/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 22026435 Supplier Azer Scientific Supplier No. PFNBF20
Only null left

Designed specifically for the collection and transport of various sizes of tissue samples

  • A traditional fixative that is buffered to a neutral pH
  • Ideal for histological applications for fixing and storing tissue specimens
  • Each container is half filled with 10 percent neutral buffered formalin
  • Packaged with our proprietary Sure-Lok closure

Compliance

Containers are 95 kPa compliant.

Specifications

Capacity 20 mL
Content And Storage 10 mL NBF 10%
Packaging Type 32 Pk., 4 Pks/Cs.
Product Type Formalin
For Use With (Application) Fixative, safe collection, preservation, transportation, and storage of specimens of various sizes
Format Container