missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azer Scientific Pre-Filled 10% Neutral Buffered Formalin
Description
- A traditional fixative that is buffered to a neutral pH
- Ideal for histological applications for fixing and storing tissue specimens
- Each container is half filled with 10 percent neutral buffered formalin
- Packaged with our proprietary Sure-Lok closure
Compliance
Containers are 95 kPa compliant.
Specifications
Specifications
| Capacity | 20 mL |
| Content And Storage | 10 mL NBF 10% |
| Packaging Type | 32 Pk., 24 Pks/Cs. |
| Product Type | Formalin |
| For Use With (Application) | Fixative, safe collection, preservation, transportation, and storage of specimens of various sizes |
| Format | Container |