missing translation for 'onlineSavingsMsg'
Learn More

Azer Scientific Pre-Filled 10% Neutral Buffered Formalin

A traditional fixative buffered to a neutral pH

Supplier:  Azer Scientific PFNBF1000

 View more versions of this product

Catalog No. 22026449


Only null left

Description

Description

  • Designed specifically for the collection and transport of various sizes of tissue samples
  • Each container is half filled with 10% neutral buffered formalin
  • Packaged with our proprietary Sure-Lok™ system

Histological applications for fixing and storing tissue specimens

Compliance

These containers are 95 kPa compliant

Specifications

Specifications

33.8 oz., pre-filled 16.9 oz.
1000 mL, pre-filled 500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions