missing translation for 'onlineSavingsMsg'
Learn More

Azer Scientific Hematology Analyzer Slide

Catalog No. 22026292
Change view
Click to view available options
Hematology analyzer slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22026292 Hematology analyzer slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22026292 Supplier Azer Scientific Supplier No. EMS100TPB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for use in automated hematology slide makers

White glass slides feature printed white bars at either end, designed for blood smearing in the hematology lab

  • Comparable to those provided by Beckman Coulter™
  • White glass ensures optimal clarity and lower auto-fluorescence
  • Finishing process creates a 100% wettable surface, creating easy and uniform blood smears
  • Two printed bars act as a spacer, reducing jamming and scratching of the slides

For use in the Coulter™ LH 780™ Hematology Analyzer LH SlideMaker™ instrument

Specifications

Material Glass
Type Hematology Analyzer Slide
Color White
For Use With (Application) Designed for use in automated hematology slide makers