Learn More
Azer Scientific Hematology Analyzer Slide with Thermal Print
Catalog No. 22026293
Change view
Click to view available options
Hematology Analyzer Slide with Thermal Print
Catalog No.
22026293 Hematology Analyzer Slide with Thermal Print
1 options
Catalog No. 22026293 Supplier Azer Scientific Supplier No. EMS100TH
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Microscope slides for hematology analyzers with thermal print areas
  • Clipped corners
  • 75 × 25 × 1 mm
  • Single frosted end
  • 72 slides/box and 20 boxes/case (1400/cs)
  • compatible with hematology analyzers w/ thermal print area
Specifications
Material Glass
For Use With (Application) hematology analyzers with thermal print areas