Learn More
Azer Scientific Choyce Clean Catch Urine Collection Kit
Used to conduct culture and sensitivity studies for determining efficacy of antibiotics
Supplier: Azer Scientific ES63321
Description
- Kit (21-100-700): contains one sterile 90mL/53mm urine collection cup and includes Azer Grip handle to prevent contamination, two castile soap towelettes and instructions in Spanish and English
- Grip Handles (21-100-701): can be purchased separately in cases of 500
Specifications
Urine Collection Kit | |
Screw Cap | |
Used to conduct culture and sensitivity studies for determining efficacy of antibiotics | |
One sterile 90 mL collection cup, grip handle, two castile soap towelettes and instructions in Spanish and English |