Learn More
Azer Scientific Choyce Clean Catch Urine Collection Kit
Used to conduct culture and sensitivity studies for determining efficacy of antibiotics
Supplier:  Azer Scientific ES64310
 View more versions of this product
Catalog No. 21100701
Only null left
Description
Description
  • Kit (21-100-700): contains one sterile 90mL/53mm urine collection cup and includes Azer Grip handle to prevent contamination, two castile soap towelettes and instructions in Spanish and English
  • Grip Handles (21-100-701): can be purchased separately in cases of 500
Specifications
Chemical Identifiers
Used to conduct culture and sensitivity studies for determining efficacy of antibiotics
Specifications
Grip Handle
Used to conduct culture and sensitivity studies for determining efficacy of antibiotics
For Urine collection kit container
Product Suggestions
Product Suggestions