missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Azelnidipine

Chemical Identifiers

CAS 123524-52-7
Chemical Name or Material Azelnidipine
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC465060010
View Documents Promotion Details
Thermo Scientific™
465060010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC465060050
View Documents Promotion Details
Thermo Scientific™
465060050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

123524-52-7
Azelnidipine

Specifications

123524-52-7
Solubility aq. soln.: insoluble aq. soln.. Other solubilities: insoluble in hexane, soluble in aceteone, dmf, methanol, ethanol and, acetonitrile
193.3°C to 195.0°C
Azelnidipine
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

Causes serious eye damage.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only