Learn More
Thermo Scientific™ Azelnidipine
Supplier: Thermo Scientific™ 465060050
| Quantity | 5 g |
|---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 123524-52-7 | |
| Azelnidipine |
Specifications
| Azelnidipine | |
| 5 g | |
| 193.3°C to 195.0°C |
| 123524-52-7 | |
| Solubility aq. soln.: insoluble aq. soln.. Other solubilities: insoluble in hexane, soluble in aceteone, dmf, methanol, ethanol and, acetonitrile |
Safety and Handling
GHS H Statement Harmful if swallowed.
Causes serious eye damage.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger