missing translation for 'onlineSavingsMsg'
Learn More

Azelaoyl Chloride 98.0+%, TCI America™

Catalog No. A056225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A056225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A056225ML Supplier TCI America Supplier No. A056225ML
Only null left

Chemical Identifiers

CAS 123-98-8
Molecular Formula C9H14Cl2O2
Molecular Weight (g/mol) 225.109
MDL Number MFCD00000767
InChI Key HGEVGSTXQGZPCL-UHFFFAOYSA-N
Synonym azelaoyl chloride, azelayl chloride, azeoloyl chloride, azelaic acid chloride, azelaic acid dichloride, azelaoylchloride, azeloylchloride, nonanedioyl chloride, acmc-209aqf
PubChem CID 67165
IUPAC Name nonanedioyl dichloride
SMILES C(CCCC(=O)Cl)CCCC(=O)Cl

Specifications

Color Yellow
Boiling Point 172°C
Quantity 25 mL
UN Number 3265
Formula Weight 225.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Azelaoyl Chloride