missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azelaic acid, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS | 123-99-9 |
| Assay Percent Range | 98% |
| Molecular Formula | C9H16O4 |
| Linear Formula | HOOC(CH2)7COOH |
| MDL Number | MFCD00004432 |
| Synonym | azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110 |
| InChI Key | BDJRBEYXGGNYIS-UHFFFAOYSA-N |
| SMILES | C(CCCC(=O)O)CCCC(=O)O |
| IUPAC Name | nonanedioic acid |
| Molecular Weight (g/mol) | 188.223 |
| Show More |
RUO – Research Use Only