missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azelaic acid, 98%
CAS: 123-99-9 | C9H16O4 | 188.223 g/mol
$48.74 - $48.74
Chemical Identifiers
| CAS | 123-99-9 |
|---|---|
| Molecular Formula | C9H16O4 |
| Molecular Weight (g/mol) | 188.223 |
| MDL Number | MFCD00004432 |
| InChI Key | BDJRBEYXGGNYIS-UHFFFAOYSA-N |
| Synonym | azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110 |
| PubChem CID | 2266 |
| ChEBI | CHEBI:48131 |
| IUPAC Name | nonanedioic acid |
| SMILES | C(CCCC(=O)O)CCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC401520250
|
Thermo Scientific Chemicals
401520250 |
25 g | Glass bottle |
Each for $48.74
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 123-99-9 | |
| 188.223 | |
| BDJRBEYXGGNYIS-UHFFFAOYSA-N | |
| 2266 | |
| nonanedioic acid |
| C9H16O4 | |
| MFCD00004432 | |
| azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110 | |
| CHEBI:48131 | |
| C(CCCC(=O)O)CCCC(=O)O |
Specifications
| 98.0°C to 103.0°C | |
| 237.0°C (20.0hPa) | |
| 98% | |
| HOOC(CH2)7COOH | |
| azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110 | |
| BDJRBEYXGGNYIS-UHFFFAOYSA-N | |
| nonanedioic acid | |
| 2266 | |
| 188.22 | |
| 25 g | |
| Authentic | |
| Slightly Crystalline Powder or Flakes |
| 123-99-9 | |
| White to Yellow | |
| C9H16O4 | |
| MFCD00004432 | |
| Solubility in water: 2.4g/L water (20°C). Other solubilities: very soluble in alcohol and ether:2.7g/100mL(15°C), soluble in organic solvents, slightly soluble in benzene | |
| C(CCCC(=O)O)CCCC(=O)O | |
| 188.223 | |
| CHEBI:48131 | |
| 98% | |
| 215°C | |
| Glass bottle | |
| Azelaic acid, 98% |
RUO – Research Use Only