missing translation for 'onlineSavingsMsg'
Learn More

Azelaic acid, 96%

Catalog No. p-7052116
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4258714 25 g
AA4258722 100 g
AA4258736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4258714 Supplier Thermo Scientific Chemicals Supplier No. 4258714
Only null left

CAS: 123-99-9 | C9H16O4 | 188.223 g/mol

Azelaic acid acts as an anti-inflammatory compound. It is also exhibits anti acne activity. It up regulates expression of ant oxidative enzymes. Azelaic acid also inhibits proliferation of melanocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-99-9
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00004432
InChI Key BDJRBEYXGGNYIS-UHFFFAOYSA-N
Synonym azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110
PubChem CID 2266
ChEBI CHEBI:48131
IUPAC Name nonanedioic acid
SMILES C(CCCC(=O)O)CCCC(=O)O

Specifications

Melting Point ∼102°C
Boiling Point 286°C (100mmHg)
Beilstein 1101094
Merck Index 14,905
Solubility Information Soluble in water (2.4mg/ml at 20°C),ethanol (5%),DMSO,methanol (70mg/ml),and alcohol.
Formula Weight 188.22
Percent Purity 96%
Density 1.225 g/mL
Quantity 25 g
Flash Point 215°C (419°F)
Physical Form Crystalline
Chemical Name or Material Azelaic acid
Show More Show Less
EINECSNumber 204-669-1
RTECSNumber CM1980000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only