missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azaperone
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Loss on Drying | 0.5% max. (60°C, 4 h) (vacuum) |
| Infrared Spectrum | Authentic |
| Quantity | 1 g |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in methanol, chloroform |
| Formula Weight | 327.4 |
| Chemical Name or Material | Azaperone |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. |
| Precautionary Statement | GHS P Statement Wash face,hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. |
| EINECSNumber | 216-715-8 |
RUO – Research Use Only