missing translation for 'onlineSavingsMsg'
Learn More

Azadibenzocyclooctyne-amine

Catalog No. AAJ65637MC
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65637MC 100 mg
AAJ65637MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65637MC Supplier Thermo Scientific Chemicals Supplier No. J65637MC

CAS: 1255942-06-3 | C18H16N2O | 276.339 g/mol

Azadibenzocyclooctyne amine is a carbonyl reactive reagent used to incorporate ADIBO into organic compounds, surfaces or particlesAzadibenzocyclooctyne amine is widely useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azadibenzocyclooctyne amine is a carbonyl reactive reagent used to incorporate ADIBO into organic compounds, surfaces or particlesAzadibenzocyclooctyne amine is widely useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

Notes
Heat sensitive. Incompatible with amines and azides.

Chemical Identifiers

CAS 1255942-06-3
Molecular Formula C18H16N2O
Molecular Weight (g/mol) 276.339
MDL Number MFCD22380759
InChI Key OCCYFTDHSHTFER-UHFFFAOYSA-N
Synonym dbco-amine, azadibenzocyclooctyne-amine, adibo-ch2 2-nh2.tfa, azadibenzocyclooctyne-ch2 2-amine.tfa, dibenzocyclooctyne-amine, for copper-free click chemistry, 5-beta-alanyl-11,12-didehydro-5,6-dihydrodibenzo b,f azocine, 3-amino-1-5-aza-3,4:7,8-dibenzocyclooct-1-yne-5-yl-1-propanone, 3-amino-1-2-azatricyclo 10.4.0.0?,? hexadeca-1 16 ,4,6,8,12,14-hexaen-10-yn-2-yl propan-1-one, 3-amino-1-2-azatricyclo 10.4.0.04,9 hexadeca-1 12 ,4 9 ,5,7,13,15-hexaen-10-yn-2-yl propan-1-one
PubChem CID 77078258
SMILES C1C2=CC=CC=C2C#CC3=CC=CC=C3N1C(=O)CCN

Specifications

Color Yellow to Orange
Quantity 100 mg
Formula Weight 276.33
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne-amine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only