Learn More
Axygen™ AxyMats™ Sealing Mats for 500uL 96 Well Plates
Catalog No. 50400309
Change view
Click to view available options
Pre-slit:
No
Yes
Sterility:
Non-sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pre-slit Sterility
50400309 No Non-sterile
14221903 Yes Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50400309 Supplier Axygen™ Supplier No. AM500ULRD
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Color Natural
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
No. per Case 50/Cs.
No. per Pack 10/Pk.
Pre-slit No
Product Type Sealing Mat
Well Shape Round
Plate Compatibility 96-well Deep Well Plate, 0.5 mL
Show More Show Less