Learn More
Axygen™ AxyMats™ Sealing Mats for 500uL 96 Well Plates
Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Specifications
| Color | White |
| Material | Silicone |
| Sterility | Non-sterile |
| No. of Wells | 96 |
| No. per Pack | 10 Mats |
| Pre-slit | Yes |
| Product Type | Slitted Microplate Sealing Mat |
| Plate Compatibility | 96-well Deep Well Plate, 0.5 mL |