missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Micropipette Tips

Catalog No. 14222911
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Round
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Tip Style
14222911 Non-sterile Automation, Round
14222912 Sterile Automation, Round
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14222911 Supplier Axygen™ Supplier No. ZT200R

Axygen™ Automation Micropipette Tips are constructed of polypropylene, and feature a variety of available options to suit your needs.

  • For Tecan™/Freedom EVO&trade, Te-Mo™, Aquarius, Zymark™/Caliper, RapidPlate™, and SciClone™ Systems
  • For 96-well, Non-ANSI and ANSI/SLAS (384) format, number 5 polypropylene racks
  • Available with or without aerosol-resistant filters
  • Available sterile/non-sterile
  • Available in standard or Maxymum Recovery™ style

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Blue
Autoclavable Autoclavable
Clean Claims RNase-free, DNase-free, Nonpyrogenic
Format Rack
Tip Style Automation, Round
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Quantity 50 Racks/Cs.
Product Type Automation Micropipette Tip
For Use With (Equipment) Tecan/Freedom EVO, Te-Mo, Aquarius, Zymark/Caliper, RapidPlate, and SciClone Systems
Material Polypropylene
Show More Show Less