missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Tissue Grinder

Catalog No. 14222358
Change view
Click to view available options
Packaging:
Individually wrapped
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
14222358 Individually wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14222358 Supplier Axygen™ Supplier No. PES15BSI
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fits 1.5 and 2.0mL centrifuge tubes

  • Durable and inert polypropylene construction
  • Designed to fit 1.5 mL and 2.0 mL microcentrifuge tubes

Specifications

Autoclavable Autoclavable
Color Blue
For Use With (Equipment) 1.5 and 2 mL microcentrifuge tube and PCR tube
Packaging Individually wrapped
Material Polypropylene
Sterility Sterile