missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Tip Refill System, Empty Racks

Catalog No. 14222439
Change view
Click to view available options
Model:
RFL-R
Packaging:
10 Racks/Pack; 5 Packs/Case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Packaging
14222439 RFL-R 10 Racks/Pack; 5 Packs/Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14222439 Supplier Axygen™ Supplier No. RFLR
Get Up to $2,500 in Products with Purchase
View all promotions

Easy-to-load refill racks for Axygen Tip Refill System. For use with P2/P10 Gilson-Style and 200μL Universal fit inserts.

Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.

  • RFL-R is suitable for both the 10 μL and 200 μL refill inserts
  • RFL-R-1200 is suitable for 1200 μL refill inserts
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip

Specifications

Color Clear
Sterility Non-sterile
For Use With (Equipment) P2/P10 Gilson-Style and 200μL Universal fit inserts
Model RFL-R
Packaging 10 Racks/Pack; 5 Packs/Case
Product Type Empty Refill Rack with Lid