missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Thin Wall PCR Tubes with Flat Cap, Clear, Nonsterile
GREENER_CHOICE

Catalog No. 14223223
Change view
Click to view available options
Volume (Metric):
0.2 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14223223 0.2 mL
14223224 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14223223 Supplier Axygen™ Supplier No. PCR02C500

Ultra-thin and consistent wall thickness

Axygen™ polypropylene thin wall PCR tubes with flat caps are designed to fit standard 96 well blocks and are available in an assortment of colors. Ultra-thin and consistent wall thickness allows precise thermal transfer and even insertion cap ensures a tight fit to prevent evaporation.
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene

Specifications

Certifications/Compliance Certified RNase-DNase-free
Color Clear
Volume (Metric) 0.2 mL
Closure Material Polypropylene
Shape Conical
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap
Format Fits standard 96 well blocks