missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Tecan™ Evo MCA96 Compatible Nested Pipet Tips
GREENER_CHOICE

Catalog No. 14223553
Change view
Click to view available options
Volume (Metric):
50 μL
200 μL
Sterility:
Non-sterile
Sterile
Color:
Gray
Orange
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Color
14223553 200 μL Sterile Gray
14223552 200 μL Non-sterile Gray
14223551 50 μL Sterile Orange
14223550 50 μL Non-sterile Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14223553 Supplier Axygen™ Supplier No. EV200NTRS
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

768 tips for use with Tecan Evo MCA96 multi-channel arm

  • Tip format: 96 tip
  • Tip volume range: 50 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
  • Compatible heads: MCA96
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene

Specifications

Volume (Metric) 200 μL
Sterility Sterile
Color Gray
Tip Style Round
Packaging Nested tip rack
Quantity 40 Racks/Cs.
For Use With (Equipment) Tecan EVO MCA96
Product Line Tecan
Material Polypropylene