missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
GREENER_CHOICE

Catalog No. 14222224
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14222224 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14222224 Supplier Axygen™ Supplier No. P2MLSQC
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For sample collection, in-vitro growth chambers and long term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
 

  • 2.2 mL deep well plate
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film

Currently experiencing high demand for this item. Posted availability date may not be current. Contact Customer Support for up-to-date availability.

Specifications

Certifications/Compliance ANSI, SLAS 1-2004
Lid Without Lid
Array 8 x 12
Color Clear
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Square
For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
No. per Case 25/Cs.
No. per Pack 5/Pk.
Temperature Range (English) -112°F
Temperature Range (Metric) -80°C
Show More Show Less