missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
GREENER_CHOICE

Catalog No. 14222356
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14222356 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14222356 Supplier Axygen™ Supplier No. PDW500C
  • ONLINE EXCLUSIVE PRICE
    $81.10
    $31.95 / Pack of 5 Each

    Save $49.15

    61.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • ONLINE EXCLUSIVE PRICE
    $811.00
    $319.50 / Case of 10 Pack

    Save $491.50

    61.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For sample collection, in-vitro growth chambers and long term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • 0.6 mL deep well plate
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • Specifications

    Certifications/Compliance ANSI, SLAS 1-2004
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Autoclavable Autoclavable
    Color Clear
    Material Polypropylene
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
    Max. RCF 4000 x g
    No. per Case 50/Cs.
    No. per Pack 5/Pk.
    Temperature Range (English) -112°F
    Temperature Range (Metric) -80°C
    Show More Show Less