missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates

Catalog No. 50400501
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
50400501 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50400501 Supplier Axygen™ Supplier No. P96450VY

For sample collection, in-vitro growth chambers and long term storage.

Available in a variety of configurations.

Specifications

Certifications/Compliance ANSI, SLAS 1-2004
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Color Yellow
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
Max. RCF 4000 x g
Temperature Range (English) -112°F
Temperature Range (Metric) -80°C
Show More Show Less