missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Slicone 96 Round Well Compression Flat Mat for PCR Microplates

Catalog No. 50400352
Change view
Click to view available options
Material:
Silicone
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
50400352 Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50400352 Supplier Axygen™ Supplier No. CMFLAT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suited to help reduce evaporation and ensure proper seal when used in conjunction with Axygen PCR Microplates and PlateMax™ sealing films

Axygen™ Compression Mats are ideally suited to help reduce evaporation and ensure a proper seal when used in conjunction with Axygen PCR Microlates and PlateMaxTM sealing films. Made of inert 100% virgin silicone, they are capable of withstanding numerous PCR cycles.

  • For use with 96/384 well and PCR microplates
  • Specifications

    Color Natural
    Material Silicone
    Sterility Non-sterile
    Autoclavable Autoclavable
    No. of Wells 96
    No. per Case 50/Cs.
    No. per Pack 10/Pk.
    Product Type Compression Mat
    Well Shape Round
    Plate Compatibility 96/384-well PCR Microplate