Learn More
Axygen™ Septa Mat for ABI multi-capillary Sequencing Instrument
Septa Mat for ABI Multi-Capillary Sequencing Instrument
Supplier:  Axygen™ AM96SEPTA3100
Catalog No. 50-400-312
Only null left
Explore available promotions
Description
Description
Axygen™ AxyMatsTM septa mat is designed to be used on ABI sequencer. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused.
  • Septa Mat for ABI Multi-Capillary Sequencing Instrument
  • For use with 96 PCR microplates
  • Specifications
    Specifications
    ABI Multi-capillary Sequencing Instruments
    Septa Mat
    Product Suggestions
    Product Suggestions