missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ PerkinElmer™ Compatible Tips

Catalog No. 14223067
Change view
Click to view available options
Volume (Metric):
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14223067 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14223067 Supplier Corning Incorporated Supplier No. PKF384RS

384-well tips for use with PerkinElmer

  • DNAse-Free, RNAse-Free, Pyrogen-Safe
  • 384-Tip format for MDT 384-30 μL head
  • Clear Polypropylene
  • Sterile
  • With Aerosol-Resistant Filters
  • Packaging: 384 tips/rack, 10 racks/pk, 5 packs/case

Specifications

Volume (Metric) 30 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Format Rack
Tip Style Round
For Use With (Equipment) PerkinElmer
Product Line PerkinElmer
Material Polypropylene