missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Multimek Robotic Tips: 50μL

Catalog No. 14222194
Change view
Click to view available options
Volume (Metric):
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14222194 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14222194 Supplier Corning Incorporated Supplier No. MT50LRS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fits Multimek Type II or CCS-style head

Robotic tip's unique diamond-polished molding technology uses no mold-releasing agents. Minimizes wastage of precious samples and reagents while handling. Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination.

  • Tip volume: 50 and 200μL
  • Tip color: Clear
  • Rack color: Red and blue

Order Info

Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

Specifications

Volume (Metric) 50 μL
Sterility Sterile
Color Clear
Rack Color Red
Clean Claims DNase, RNase, and Pyrogen free
For Use With (Application) Unique diamond polished molding technology use no mold-releasing agents, minimizes wastage of precious samples and reagents while handling, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
Format Rack
Tip Style Low Retention
For Use With (Equipment) Beckman Coulter Multimek
Material Polypropylene