missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Microplate Sealing Film and Tapes

Catalog No. 50400316
Change view
Click to view available options
Material:
Aluminum
Polyester
Polyolefin
Rayon
Product Type:
AxySeal Sealing Film
Breathable Sealing Film
CyclerSeal Sealing Film
Sealing Film
UltraClear Sealing Film
Dimensions (L x W):
133.3 x 79.2 mm
Not Available
135.5 x 80 mm
146 x 79.2 mm
146 x 79.6 mm
153 x 82.3 mm
Sterility:
Non-sterile
Sterile
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type Dimensions (L x W) Sterility
50400316 Rayon Breathable Sealing Film 153 x 82.3 mm Non-sterile
14222044 Rayon Breathable Sealing Film 153 x 82.3 mm Sterile
14222342 Aluminum Sealing Film 146 x 79.2 mm Non-sterile
14222343 Aluminum Sealing Film 133.3 x 79.2 mm Non-sterile
14222346 Polyester AxySeal Sealing Film 146 x 79.6 mm Sterile
14222344 Polyester AxySeal Sealing Film 146 x 79.6 mm Non-sterile
14222347 Polyester CyclerSeal Sealing Film 135.5 x 80 mm Non-sterile
14222873 Polyolefin UltraClear Sealing Film Not Available Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 50400316 Supplier Axygen™ Supplier No. BF400

Offer great economy for disposable ease.

For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.

Breathable

  • Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
  • Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
Aluminum
  • Provides exceptional seal on multi-well plates with varied temperatures and reagents
  • With compression mats, films and tapes seal during thermocycling
  • Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
  • Pierceable, PCR-AS-200
CyclerSeal
  • Exceptional seal across entire plate safeguards samples during transport or storage
  • When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
AxySeal
  • Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
  • Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
  • Available in both nonsterile and sterile packaging

Specifications

Material Rayon
Sterility Non-sterile
Length (English) 6.02 in.
Length (Metric) 153 mm
No. per Case 1000/Cs.
No. per Pack 100/Pk.
Dimensions (L x W) 153 x 82.3 mm
Width (English) 3.24 in.
Width (Metric) 82.3 mm
For Use With (Application) Designed for applications ranging from PCR and real-time PCR to ELISA and cell culture, allow effective gas exchange for cellular and bacterial cultivation, while preventing contamination
Product Type Breathable Sealing Film
Thickness 50 μm
Temperature Range (Metric) -20°C to +80°C
Show More Show Less