missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Microplate Sealing Film and Tapes
Description
For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.
Breathable
- Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
- Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
- Provides exceptional seal on multi-well plates with varied temperatures and reagents
- With compression mats, films and tapes seal during thermocycling
- Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
- Pierceable, PCR-AS-200
- Exceptional seal across entire plate safeguards samples during transport or storage
- When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
- Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
- Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
- Available in both nonsterile and sterile packaging
Specifications
Specifications
| Material | Polyester |
| Sterility | Non-sterile |
| Length (English) | 5.33 in. |
| Length (Metric) | 135.5 mm |
| No. per Case | 500/Cs. |
| No. per Pack | 100/Pk. |
| Dimensions (L x W) | 135.5 x 80 mm |
| Width (English) | 3.14 in. |
| Width (Metric) | 80 mm |
| For Use With (Application) | Designed for applications ranging from PCR and real-time PCR to ELISA and cell culture |
| Show More |