missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Microplate Sealing Film and Tapes

Catalog No. 14222343
Change view
Click to view available options
Material:
Aluminum
Polyester
Polyolefin
Rayon
Product Type:
AxySeal Sealing Film
Breathable Sealing Film
CyclerSeal Sealing Film
Sealing Film
UltraClear Sealing Film
Dimensions (L x W):
133.3 x 79.2 mm
Not Available
135.5 x 80 mm
146 x 79.2 mm
146 x 79.6 mm
153 x 82.3 mm
Sterility:
Non-sterile
Sterile
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type Dimensions (L x W) Sterility
14222343 Aluminum Sealing Film 133.3 x 79.2 mm Non-sterile
50400316 Rayon Breathable Sealing Film 153 x 82.3 mm Non-sterile
14222044 Rayon Breathable Sealing Film 153 x 82.3 mm Sterile
14222342 Aluminum Sealing Film 146 x 79.2 mm Non-sterile
14222346 Polyester AxySeal Sealing Film 146 x 79.6 mm Sterile
14222344 Polyester AxySeal Sealing Film 146 x 79.6 mm Non-sterile
14222347 Polyester CyclerSeal Sealing Film 135.5 x 80 mm Non-sterile
14222873 Polyolefin UltraClear Sealing Film Not Available Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14222343 Supplier Axygen™ Supplier No. PCRAS600
  • ONLINE EXCLUSIVE PRICE
    $373.75
    $228.62 / Pack of 100 Each

    Save $145.13

    39.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • ONLINE EXCLUSIVE PRICE
    $1,868.75
    $1,143.10 / Case of 5 Pack

    Save $725.65

    39.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Offer great economy for disposable ease.

For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. 

Aluminum

  • Provides exceptional seal on multi-well plates with varied temperatures and reagents
  • With compression mats, films and tapes seal during thermocycling
  • Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
  • Pierceable, PCR-AS-200

 

Specifications

Material Aluminum
Sterility Non-sterile
Length (English) 5.24 in.
Length (Metric) 133.3 mm
No. per Case 500/Cs.
No. per Pack 100/Pk.
Dimensions (L x W) 133.3 x 79.2 mm
Width (English) 3.11 in.
Width (Metric) 79.2 mm
For Use With (Application) Designed for applications ranging from PCR and real-time PCR to ELISA and cell culture, ideal for light-sensitive samples
Product Type Sealing Film
Thickness 50 μm
Temperature Range (Metric) -80°C to +104°C
Show More Show Less