missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ LightCycler 480 Compatible PCR Plates, 96-well
GREENER_CHOICE

Catalog No. 14223389
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14223389 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14223389 Supplier Axygen™ Supplier No. PCR96LC480CNF
Only null left

96-well plate is designed to fit the Roche LightCycler™ 480

  • Certified RNase- and DNase-free
  • Nonsterile
  • Large print alphanumeric grid for ease of well identification

Specifications

Barcode No Barcode
Certifications/Compliance RNase, DNase-free
Format Standard
Array 8 x 12
Color Clear
For Use With (Equipment) For Roche light cycler 480
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) RT-PCR
Includes Printed alphanumeric grid
No. per Case 50/Cs.
No. per Pack 10/Pk.
Show More Show Less