missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Horizontal Gel Box, 10 cm

Catalog No. 14223901
Change view
Click to view available options
Dimensions (L x W):
12.5 x 22 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
14223901 12.5 x 22 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14223901 Supplier Axygen™ Supplier No. HGB10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Axygen HGB-10 is a compact 10cm wide gel box capable of running up to 100 samples depending on configuration. System is ideal for quick sample-checks, particularly following restriction digestion during cloning. Comes with two different size UV transparent gel trays providing flexibility and value.

Axygen horizontal gel boxes are high-quality, full-featured units designed for exceptional durability and efficiency. The gel boxes feature leak-free external gel casting, which allows you to cast one gel outside of the box while another gel is running inside. The units are available in four different sizes, and each unit comes with two sizes of gel trays. In addition, they have easy to replace electrodes that are recessed to reduce potential damage. These gel boxes are an excellent value that will provide years of reliable service.



Features
  • Flexibility – Each unit comes with 2 sizes of gel trays to optimize applications
  • External gel casting to gel box allows for running gels while casting the next one
  • Ease of use – the Axygen contrast enhancer makes loading gels easier
  • Rugged injection molding is supported by a 3-year warranty
  • Larger units come with recirculation ports allowing for buffer exchange

Four sizes of horizontal tanks. Each comes with 2 sizes of gel trays which allow for external gel casting. Three larger sizes come with recirculation ports for use as needed.

Specifications

Certifications/Compliance CE certified
Type Horizontal Gel Box
No. per Case 1/Cs.
No. per Pack 1/Pk.
Volume (Metric) Buffer 300 mL
Dimensions (L x W) 12.5 x 22 cm
Width (Metric) 10 cm