missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Gel Tips, Round

Catalog No. 14222809
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14222809 Non-sterile
14222810 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14222809 Supplier Axygen™ Supplier No. TGL200RD57R

Universal fit for research grade pipetters

Axygen™ Gel Loading Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brand of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. Available in either flat or round versions, with different outer diameters for different gel thickness.


  • Designed for removing bands from agarose gel
  • One handed operation
  • Push button gel release
  • Ejector button tip release
  • Avoid cross contamination
  • Tips fit standard 200 μL pipettors
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Nonsterile
  • Specifications

    Volume (Metric) 0.5 to 200 μL
    Filtered Non-filtered
    Sterility Non-sterile
    Color Clear
    Autoclavable Autoclavable
    Certifications/Compliance ISO 13485
    Clean Claims DNase, RNase Free
    For Use With (Application) Agarose Gel Band Removal
    Diameter (Metric) Outer 0.57 mm
    Format Rack
    Tip Style Gel Loading, Round
    Packaging 200 Tips/Rack, 2 Racks/Cs
    Quantity 200/Pk. (400/Cs.)
    For Use With (Equipment) Universal Fit
    Graduated Yes
    Material Polypropylene
    Shape Round
    Graduations 2, 5μL
    Show More Show Less