Learn More
Axygen™ Gel Tips, Flat
Catalog No. 14222807
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14222807 0.5 to 200 μL
14222801 0.1 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14222807 Supplier Axygen™ Supplier No. TGL200FT37R
Universal fit for research grade pipetters
Axygen™ Gel Loading Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brand of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. Available in either flat or round versions, with different outer diameters for different gel thickness.
  • Designed for removing bands from agarose gel
  • One handed operation
  • Push button gel release
  • Ejector button tip release
  • Avoid cross contamination
  • Tips fit standard 1000 μL pipettors
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Nonsterile
  • OD value: 0.17mm
  • Specifications
    Volume (Metric) 0.5 to 200 μL
    Sterility Non-sterile
    Color Clear
    Thickness 0.4 mm
    Clean Claims DNase, RNase Free
    Tip Style Gel Loading
    Packaging 200 Tips/Rack, 2 Racks/Cs
    Graduated Yes
    Material Polypropylene
    Shape Flat
    Graduations 2, 5μL
    Show More Show Less