missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ FDSS™ Robotic Tips
GREENER_CHOICE

Catalog No. 14223303
Change view
Click to view available options
For Use With (Equipment):
Hamamatsu FDSS 7000 System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14223303 Hamamatsu FDSS 7000 System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14223303 Supplier Axygen™ Supplier No. FDSS1536BKR
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Hamamatsu FDSS series systems

  • For 384-Well plates
  • Tip color: Black
  • Capacity: 50μL
  • Packaging: 384 Tips/rack; 10 Racks/unit; 5 Units/Cs.

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Black
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Hamamatsu FDSS 7000 System
Material Polypropylene
Show More Show Less