missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Eppendorf™ Compatible Pipet Tips

Catalog No. 14223383
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Eppendorf
Low Retention
Packaging:
Bulk
Racked
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Tip Style Packaging
14223383 Non-sterile Eppendorf Racked
14223382 Sterile Low Retention Racked
14223381 Non-sterile Low Retention Racked
14223379 Non-sterile Eppendorf Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14223383 Supplier Corning Incorporated Supplier No. TE1004BR

Compatible with Eppendorf and Biohit pipets

  • Available in sterile or nonsterile forms
  • e-Beam irradiated to prevent product degradation
  • Maxymum recovery tips allows complete sample retrieval and greater tip-to-tip precision
  • Graduation marks at 110 μL and 500 μL, allows visual confirmation of pipettor draw volume
  • Volume: 1000 μL
  • Color: Blue

Order Info

  • Bulk packed: 500 Tips/Pack. 10 Packs/Cs.
  • Racked: 96 Tips/Rack, 12 Racks/Pack, 3 Packs/Cs.

Specifications

Volume (Metric) 1000 μL
Sterility Non-sterile
Color Blue
Format Rack
Tip Style Eppendorf
Packaging Racked
For Use With (Equipment) Eppendorf
Graduated Yes
Graduations 110 and 500μL