Learn More
Axygen™ Caps for PCR Tubes
8-Strip domed or flat strip caps for Axygen PCR tubes
Supplier: Axygen™ PCR2CPRTC
Description
Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.
Specifications
| RNase, DNase, pyrogen-free | |
| MaxyClear | |
| 125/Pk. | |
| 8-strips PCR tubes and 96-well PCR microplates and also for fluorescent, luminescent and real-time PCR | |
| Polypropylene | |
| Strip |
| Clear | |
| 0.2 mL | |
| Polypropylene | |
| Non-sterile | |
| Flat Cap | |
| 8-Strip Flat PCR Tube Cap |