missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Caps for PCR Tubes
GREENER_CHOICE

Catalog No. 14222281
Change view
Click to view available options
Color:
Assorted
Clear
MaxyClear
MaxyClear, w/writing surface
Closure Type:
Domed Cap
Flat Cap
Product Type:
8-Strip Domed PCR Tube Cap
8-Strip Flat PCR Tube Cap
8-Strip Flat PCR Tube Cap with Writing Surface
PCR Caps
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type Product Type
14222281 MaxyClear, w/writing surface Flat Cap 8-Strip Flat PCR Tube Cap with Writing Surface
14222265 Clear Domed Cap 8-Strip Domed PCR Tube Cap
14222263 Assorted Domed Cap 8-Strip Domed PCR Tube Cap
14222302 MaxyClear Flat Cap 8-Strip Flat PCR Tube Cap
14223222 Clear Domed Cap PCR Caps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14222281 Supplier Axygen™ Supplier No. PCR02FCPC
  • ONLINE EXCLUSIVE PRICE
    $88.21
    $38.83 / Pack of 125 Each

    Save $49.38

    56.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • ONLINE EXCLUSIVE PRICE
    $882.10
    $388.30 / Case of 10 Pack

    Save $493.80

    56.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8-Strip domed or flat strip caps for Axygen PCR tubes

Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.

Specifications

Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Clear
Color MaxyClear, w/writing surface
Volume (Metric) 0.2 mL
Quantity 1250/Cs.
Closure Material Polypropylene
For Use With (Equipment) 8-strips PCR tubes and 96-well PCR microplates
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap
Format Strip
Product Type 8-Strip Flat PCR Tube Cap with Writing Surface
Show More Show Less