Learn More
Axygen™ AxyMats™ Pierceable Sealing Mats for 24 Well Deep Well Plates
Description
- Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications.
- AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved.
- They can be reused following a bleach wash and ethanol rinse protocol.
- Mfr. No. AM-24-SQ
Specifications
Specifications
| Material | Silicone |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| No. of Wells | 24 |
| No. per Case | 50/Cs. |
| No. per Pack | 10/Pk. |
| Product Type | Sealing Mat |
| Well Shape | Square |
| Plate Compatibility | 24-well Deep Well Plate with Square Wells |