missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Tips for STARLine, Clear, CO-RE Style Liquid Level Sensing Tip, Sterile
GREENER_CHOICE

Catalog No. 14224008
Change view
Click to view available options
Volume (Metric):
10 μL
50 μL
300 μL
For Use With (Equipment):
Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus
Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Equipment)
14224008 300 μL Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus
14224006 10 μL Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus
14224007 50 μL Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14224008 Supplier Axygen™ Supplier No. HT300CBKSTKS
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For complete compatibility and a consistent fit.

Axygen brand automation tips are engineered to be compatible with original equipment manufacturers' tips, ensuring complete compatibility and a consistent fit. One of the widest product offerings combined with our engineering expertise stringent QA/QC enables you to get optimum performance no matter which automated liquid handler you are working with.
  • Compatible with Hamilton™ Microlab™ STAR, STARlet, STARplus and Nimbus™
  • Conductive black polypropylene
  • Certified DNase-/RNase-free, nonpyrogenic
  • With or without aerosol-resistant filters
  • Nonsterile or sterile

Specifications

Volume (Metric) 300 μL
Filtered Non-filtered
Sterility Sterile
Color Black
Clean Claims DNase, RNase Free
Format Refill
Tip Style Round
For Use With (Equipment) Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus
Material Polypropylene