Learn More
Axygen™ Automation Tips for STARLine, Clear, CO-RE Style Liquid Level Sensing Tip, Sterile
For complete compatibility and a consistent fit.
Supplier: Axygen™ HT50CBKSTKS
Description
Axygen brand automation tips are engineered to be compatible with original equipment manufacturers' tips, ensuring complete compatibility and a consistent fit. One of the widest product offerings combined with our engineering expertise stringent QA/QC enables you to get optimum performance no matter which automated liquid handler you are working with.- Compatible with Hamilton™ Microlab™ STAR, STARlet, STARplus and Nimbus™
- Conductive black polypropylene
- Certified DNase-/RNase-free, nonpyrogenic
- With or without aerosol-resistant filters
- Nonsterile or sterile
Specifications
| 50 μL | |
| Sterile | |
| DNase, RNase Free | |
| Round | |
| Polypropylene |
| Non-filtered | |
| Black | |
| Refill | |
| Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus |