missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-well PCR Microplates
GREENER_CHOICE

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.

Supplier:  Axygen™ PCR96M2HSC

 View more versions of this product

Catalog No. 14222340

Only null left

Description

Description

Axygen™ 96 well elevated skirt PCR microplate is designed to fit ABI Real Time PCR thermal cyclers and sequencers. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • 96-well Semi-Skirted PCR Plate
  • Single Notch Coner
  • Application/Compatibility: Standard thermal cycling, ABI Real time PCR 7300, 7500, 7900, qPCR and ABI Sequencing
  • Specifications

    Chemical Identifiers

    No Barcode
    8 x 12
    200 μL
    Non-sterile
    RNase-, DNase-free
    Polypropylene
    96
    PCR thermal transfer, RT-PCR applications

    Specifications

    No Barcode
    Single Notch Corner
    8 x 12
    ABI PCR 2700, 2720, 9700, 9800, 7300, 7500, 7900 and ABI sequencing units
    Polypropylene
    96
    Round
    50/Cs.
    RNase-, DNase-free
    Without Lid
    Clear
    Half
    200 μL
    Non-sterile
    PCR thermal transfer, RT-PCR applications
    10/Pk.
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions