missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-Well PCR Microplates
GREENER_CHOICE

Numéro de catalogue. 14221902
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Color
14221902 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 14221902 Fournisseur Axygen™ Code fournisseur PCR96CS
Il en reste null

PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications.

PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications. A variety of colors are offered to help organize multi-step PCR processes.

 

  • 96-well PCR microplates
  • Working volume is 300μL
  • Available in a variety of colors
  • Sold 10 plates/unit; 5 units/Cs.

 

Spécifications

Certifications/Compliance RNase, DNase-free
Format Standard
Array 8 x 12
Color Clear
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
For Use With (Application) PCR thermal transfer, RT-PCR applications
Well Color Clear