missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-well Low Profile Bar Coded PCR Microplates
GREENER_CHOICE

Catalog No. 14224000
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14224000 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14224000 Supplier Axygen™ Supplier No. PCR96LPABCPBC

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications. The low profile design results in faster and more efficient PCR reactions.

  • Low profile; Optimized for fast PCR
  • Certified RNase-/DNase-free
  • Nonpyrogenic, nonsterile
  • Bar coded for traceability and identification
  • Sealing options: Axymats, heat sealing, adhesive films and foils, flat and domed cap strips

Specifications

Barcode Includes Barcode
Certifications/Compliance RNase-/DNase-free
Format Standard
Array 8 x 12
Color Clear
For Use With (Equipment) ABI thermal cyclers, ABI Fast PCR 7500, 7900 9800, Veriti and StepOne
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
For Use With (Application) PCR thermal transfer, RT-PCR applications