missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-well Segmented PCR Microplates
GREENER_CHOICE

Catalog No. 14222341
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14222341 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14222341 Supplier Axygen™ Supplier No. PCR96SGC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pre-scored to break apart into four 24 well sections. Corning™ Axygen™ 96-wewll Segmented PCR Microplates are a versatile choice for thermal cycling.

Axygen™ 96 well segmented PCR microplate is designed to fit standard 96 well blocks. Plate is pre-scored to break apart into four 24 well sections. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • 96-well Segmented PCR Plate
  • Application/Compatibility: Standard thermal cycling, Beckman CEQ Sequencers
  • Specifications

    Barcode No Barcode
    Certifications/Compliance RNase, DNase-free
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color Clear
    Product Type PCR Plate
    For Use With (Equipment) Beckman CEQ Sequencers
    Material Polypropylene
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) PCR thermal transfer, RT-PCR applications
    Max. RCF 4000 x g
    No. per Case 50/Cs.
    No. per Pack 10/Pk.
    Show More Show Less