missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-Well Low Profile PCR Microplates
GREENER_CHOICE

Catalog No. 14222338
Change view
Click to view available options
Color:
Clear
White
Skirt Style:
Half
Non-skirt
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Skirt Style
14222338 Clear Half
14222949 White Half
14222339 Clear Non-skirt
14222951 White Non-skirt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14222338 Supplier Axygen™ Supplier No. PCR96LPABC
Only null left

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications. The low profile design results in faster and more efficient PCR reactions.

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications. The

Axygen™ 96 well low profile PCR microplate is designed to fit ABI Real Time PCR thermal cyclers and sequencers. Low profile design results in faster and more efficient PCR reactions. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • 96-Well Semi-Skirted Low Profile PCR Plate
  • Optimized for Fast PCR
  • Designed for ABI Thermal Cyclers
  • Application/Compatibility: ABI Fast PCR 7500, 7900 9800, Veriti and StepOne
  • Specifications

    Barcode No Barcode
    Certifications/Compliance RNase-/DNase-free
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Packaging 100 Cases, 25 Pk.
    Product Line Axygen
    Color Clear
    Product Type Standard top
    For Use With (Equipment) ABI thermal cyclers, ABI Fast PCR 7500, 7900 9800, Veriti and StepOne
    Skirt Style Half
    Material Polypropylene
    Volume (Metric) Well 100 μL
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) PCR thermal transfer, RT-PCR applications
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Show More Show Less